Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4′-Methylacetoacetanilide 98.0+%, TCI America™

Click to view available options
Quantity:
25g
500g
2415-85-2
C11H13NO2
191.23
MFCD00043931
MJGLMEMIYDUEHA-UHFFFAOYSA-N
N-Acetoacetyl-4-methylaniline, N-Acetoacetyl-p-toluidine
17015
N-(4-methylphenyl)-3-oxobutanamide
CC1=CC=C(C=C1)NC(=O)CC(=O)C
2415-85-2
C11H13NO2
191.23
MFCD00043931
MJGLMEMIYDUEHA-UHFFFAOYSA-N
N-Acetoacetyl-4-methylaniline, N-Acetoacetyl-p-toluidine
17015
N-(4-methylphenyl)-3-oxobutanamide
CC1=CC=C(C=C1)NC(=O)CC(=O)C
Quantity 500g
Melting Point 95°C
Color White-Yellow
UN Number 2811
Formula Weight 191.23
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (HPLC,N)
Chemical Name or Material 4′-Methylacetoacetanilide

EINECSNumber : (3)-0204

TSCA : Yes