Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Methylamino)phenol Sulfate, 99.0 -101.5%, Spectrum™

Click to view available options
Quantity:
25 g
125 g
500 g
Packaging:
Amber Glass Bottle
55-55-0
55-55-0
Packaging Amber Glass Bottle
Residue after Ignition 0.001
Quantity 500 g
CAS Max % 1
Percent Purity 99.0 to 101.5%
Grade Reagent

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.