Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Methylamino)phenylboronic acid pinacol ester, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
845870-55-5
C13H20BNO2
233.12
WWGNYCWKFZIQQS-UHFFFAOYSA-N
n-methyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline,4-n-methylamino phenylboronic acid, pinacol ester,4-n-methylamino phenylboronic acid pinacol ester,methyl-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-phenyl-amine,4-n-methylamino phenylboronic acid,pinacol ester,4-methylamino phenylboronic acid pinacol ester,4-methylamino phenyl boronic acid pinacol ester,n-methyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl aniline,benzenamine, n-methyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
11957892
N-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)NC
845870-55-5
C13H20BNO2
233.12
WWGNYCWKFZIQQS-UHFFFAOYSA-N
n-methyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline,4-n-methylamino phenylboronic acid, pinacol ester,4-n-methylamino phenylboronic acid pinacol ester,methyl-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-phenyl-amine,4-n-methylamino phenylboronic acid,pinacol ester,4-methylamino phenylboronic acid pinacol ester,4-methylamino phenyl boronic acid pinacol ester,n-methyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl aniline,benzenamine, n-methyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
11957892
N-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)NC
Quantity 5g
Melting Point 65.0°C to 66.0°C
Packaging Glass bottle
Assay Percent Range 96% min. (GC)
Formula Weight 233.12
Percent Purity 97%
Chemical Name or Material 4-(Methylamino)phenylboronic acid pinacol ester

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning