Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methylaminophenol sulfate, 99%, extra pure, ACROS Organics™

Click to view available options
Quantity:
5g
100g
500g
2.5kg
55-55-0
C14H18N2O2·H2SO4
344.38
MFCD00013140
Metol
55-55-0
C14H18N2O2·H2SO4
344.38
MFCD00013140
Metol

4-Methylaminophenol sulfate, 99%, C14H18N2O2.H2SO4, CAS Number-55-55-0, Metol, 500g, 13, 441, 100.0, 98.5, Beige to White or Gray, Decomposition Information: >260 deg.C, 1.5770g/mL, 200-237-1, 344.38, Extra Pure, GHS Signal Word: Warning

4-Methylaminophenol sulfate, 99%, C14H18N2O2.H2SO4, CAS Number-55-55-0, Metol, 500g, 13, 441, 100.0, 98.5, Beige to White or Gray, Decomposition Information: >260 deg.C, 1.5770g/mL, 200-237-1, 344.38, Extra Pure, GHS Signal Word: Warning

Residue after Ignition 0.15% max
Melting Point 260.0°C
Decomposition Information Decomposition Information: >260°C
Color Beige to White or Gray
Packaging Glass Bottle
Loss on Drying 0.3% max.
Infrared Spectrum Authentic
Ignition Residue 0.15% max.
Density 1.5770g/mL
Specific Gravity 1.577
Quantity 5g
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula (CH3NHC6H4OH)2·H2SO4
Beilstein 13, 441
Merck Index 15, 6091
Solubility Information Solubility in water: 50g/L (20°C). Other solubilities: soluble in 20 parts cold water,soluble in 6 parts boiling water,slightly soluble in alcohol,insoluble in ether
Formula Weight 344.38
Physical Form Crystals or Crystalline Powder
Percent Purity 99%
Grade Extra Pure
Chemical Name or Material 4-Methylaminophenol sulfate, 99%
Show More Show Less

GHS H Statement:
Very toxic to aquatic life with long lasting effects.
May cause an allergic skin reaction.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.

Emergency Overview
Harmful if swallowed. May cause eye, skin, and respiratory tract irritation . May cause an allergic skin reaction. Danger of serious damage to health by prolonged exposure if swallowed. Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment.
NFPA
Health: 2
Flammability: 1
Instability: 1

GHS Signal Word: Warning

EINECSNumber : 200-237-1

RTECSNumber : SL8650000

TSCA : TSCA