Alfa Aesar™ 4-Methylanisole, 99%

Manufacturer: Alfa Aesar™ A11572AE

Quantity 100mL

 View more versions of this product

Catalog No. AAA11572AE

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 104-93-8
    Molecular Formula C8H10O
    Formula Weight 122.17
    MDL Number MFCD00008413
    UN Number UN3271
    Beilstein 1237336
    Synonym p-Cresyl methyl ether; 4-Methoxytoluene
    Chemical Name or Material 4-Methylanisole

    Specifications

    Formula Weight 122.17
    Health Hazard 3 GHS P Statement
    P261-P280h
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/clothing.
    TSCA TSCA
    MDL Number MFCD00008413
    CAS 104-93-8
    Odor Aromatic
    Beilstein 1237336
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ETHERS, N.O.S.
    Flash Point 58°C (136°F)
    Quantity 100mL
    Molecular Formula C8H10O
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H226-H302-H315
    Flammable liquid and vapor.
    Harmful if swallowed.
    Causes skin irritation.
    RTECS Number BZ8780000
    Boiling Point 174° to 175°C
    Melting Point −50°C
    UN Number UN3271
    Synonym p-Cresyl methyl ether; 4-Methoxytoluene
    Chemical Name or Material 4-Methylanisole
    Assay 99%

    GHS H Statement
    H226-H302-H315
    Flammable liquid and vapor.
    Harmful if swallowed.
    Causes skin irritation.

    GHS P Statement
    P261-P280h
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/clothing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling
  • Promotions

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit