Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Methylbenzamidoxime, 97%

Manufacturer:  Alfa Aesar™ H5170406

Quantity 5g

 View more versions of this product

Catalog No. AAH5170406


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

19227-13-5
150.18
N-Hydroxy-4-methylbenzamidine; N-Hydroxy-4-methylbenzenecarboximidamide
C8H10N2O
MFCD00019952
4-Methylbenzamidoxime

Specifications

135° to 138°C
19227-13-5
150.18
N-Hydroxy-4-methylbenzamidine; N-Hydroxy-4-methylbenzenecarboximidamide
97%
5g
C8H10N2O
MFCD00019952
4-Methylbenzamidoxime

Safety and Handling

SDS & Safety Documents

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning

RTECSNumber : XS4870000

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit