Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Methylbenzenethiol sodium salt, 98%

Manufacturer:  Alfa Aesar™ H5509514

Quantity 25g

 View more versions of this product

Catalog No. AAH5509514


  Check Availability
Add to cart

Specifications

Description

Applications
4-Methylbenzenethiol sodium salt is used as a pharmaceutical intermediate.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Description & Specifications

Chemical Identifiers

10486-08-5
146.18
MFCD00066208
4-Methylbenzenethiol sodium salt

Specifications

White to off-white
25g
Powder
MFCD00066208
98%
>300°C
10486-08-5
146.18
4-Methylbenzenethiol sodium salt
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit