Alfa Aesar™ 4-Methylbenzenethiol sodium salt, 98%

Manufacturer: Alfa Aesar™ H5509503

Quantity 1g

 View more versions of this product

Catalog No. AAH5509503

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 10486-08-5
    Formula Weight 146.18
    MDL Number MFCD00066208
    Chemical Name or Material 4-Methylbenzenethiol sodium salt

    Specifications

    Formula Weight 146.18
    CAS 10486-08-5
    Color White to off-white
    Melting Point >300°C
    Quantity 1g
    Physical Form Powder
    MDL Number MFCD00066208
    Chemical Name or Material 4-Methylbenzenethiol sodium salt
    Assay 98%

    Description

    Applications
    4-Methylbenzenethiol sodium salt is used as a pharmaceutical intermediate.

    Notes
    Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit