Alfa Aesar™ 4-Methylbenzyl acetate, 98+%

Manufacturer: Alfa Aesar™ B2231818

Quantity 50g

 View more versions of this product

Catalog No. AAB2231818

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2216-45-7
    Molecular Formula C10H12O2
    Formula Weight 164.2
    MDL Number MFCD00017234
    Beilstein 2046163
    Chemical Name or Material 4-Methylbenzyl acetate

    Specifications

    Formula Weight 164.2
    TSCA TSCA
    Molecular Formula C10H12O2
    MDL Number MFCD00017234
    CAS 2216-45-7
    Beilstein 2046163
    Boiling Point ≈100°C (10mmHg)
    Quantity 50g
    RTECS Number DA4940000
    Chemical Name or Material 4-Methylbenzyl acetate
    Assay ≥98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit