Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Methylbenzyl acetate, 98+%

Click to view available options
Quantity:
10g
50g
250g
2216-45-7
C10H12O2
164.204
MFCD00017234
WDCUPFMSLUIQBH-UHFFFAOYSA-N
4-methylbenzyl acetate,p-methylbenzyl acetate,p-tolubenzyl acetate,p-xylyl acetate,4-methylbenzyl ethanoate,p-acetoxymethyltoluene,4-methylphenyl methyl acetate,benzenemethanol, 4-methyl-, acetate,p-methylbenzyl alcohol acetate,4-methylbenzenemethanol acetate
75187
(4-methylphenyl)methyl acetate
CC1=CC=C(C=C1)COC(=O)C
2216-45-7
C10H12O2
164.204
MFCD00017234
WDCUPFMSLUIQBH-UHFFFAOYSA-N
4-methylbenzyl acetate,p-methylbenzyl acetate,p-tolubenzyl acetate,p-xylyl acetate,4-methylbenzyl ethanoate,p-acetoxymethyltoluene,4-methylphenyl methyl acetate,benzenemethanol, 4-methyl-, acetate,p-methylbenzyl alcohol acetate,4-methylbenzenemethanol acetate
75187
(4-methylphenyl)methyl acetate
CC1=CC=C(C=C1)COC(=O)C
Quantity 10g
Boiling Point ~100°C (10mmHg)
Beilstein 2046163
Formula Weight 164.2
Percent Purity ≥98%
Chemical Name or Material 4-Methylbenzyl acetate

RTECSNumber : DA4940000

TSCA : TSCA