Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methylbenzyl cyanide, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
2947-61-7
C9H9N
131.18
MFCD00001922
RNHKXHKUKJXLAU-UHFFFAOYSA-N
4-methylbenzyl cyanide,4-methylphenylacetonitrile,p-tolylacetonitrile,p-methylbenzyl cyanide,2-4-methylphenyl acetonitrile,benzeneacetonitrile, 4-methyl,4-methylphenyl acetonitrile,4-methylbenzeneacetonitrile,4-methylbenzylcyanide,2-p-tolylacetonitrile
76280
2-(4-methylphenyl)acetonitrile
CC1=CC=C(C=C1)CC#N
2947-61-7
C9H9N
131.18
MFCD00001922
RNHKXHKUKJXLAU-UHFFFAOYSA-N
4-methylbenzyl cyanide,4-methylphenylacetonitrile,p-tolylacetonitrile,p-methylbenzyl cyanide,2-4-methylphenyl acetonitrile,benzeneacetonitrile, 4-methyl,4-methylphenyl acetonitrile,4-methylbenzeneacetonitrile,4-methylbenzylcyanide,2-p-tolylacetonitrile
76280
2-(4-methylphenyl)acetonitrile
CC1=CC=C(C=C1)CC#N
Melting Point 18°C
Flash Point 106°C
Boiling Point 242°C to 243°C
Specific Gravity 0.99
Density 0.9900g/mL
Refractive Index 1.5170 to 1.5195
Quantity 25g
Color Colorless to Yellow
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula CH3C6H4CH2CN
Beilstein 09, 530
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol,ethyl ether,benzene
Formula Weight 131.18
Physical Form Liquid
Percent Purity ≥97%
Chemical Name or Material 4-Methylbenzyl cyanide
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning