Alfa Aesar™ 4-Methylbenzyl mercaptan, 97%

Manufacturer: Alfa Aesar™ A1046722

Quantity 100g

 View more versions of this product

Catalog No. AAA1046722

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 4498-99-1
    Molecular Formula C8H10S
    Formula Weight 138.23
    MDL Number MFCD00039652
    UN Number UN2810
    Beilstein 2411317
    Synonym 4-Methyl-alpha-toluenethiol
    Chemical Name or Material 4-Methylbenzyl mercaptan

    Specifications

    Formula Weight 138.23
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 4498-99-1
    Beilstein 2411317
    Health Hazard 1 Danger
    Boiling Point 116° to 118°C (35mmHg)
    Odor Stench
    Molecular Formula C8H10S
    MDL Number MFCD00039652
    UN Number UN2810
    Synonym 4-Methyl-alpha-toluenethiol
    Quantity 100g
    Health Hazard 2 GHS H Statement
    H331-H302-H315-H319-H335
    Toxic if inhaled.
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
    Chemical Name or Material 4-Methylbenzyl mercaptan
    Assay 97%

    Description

    Applications
    4-Methylbenzyl mercaptan is used as pharmaceutical intermediates.

    Solubility
    Sparingly Soluble.

    Notes
    Air Sensitive. Store away from air and oxidizing agents.

    GHS H Statement
    H331-H302-H315-H319-H335
    Toxic if inhaled.
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit