Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methylbenzylamine 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
104-84-7
C8H11N
121.18
MFCD00008123
HMTSWYPNXFHGEP-UHFFFAOYSA-N
4-methylbenzylamine,benzenemethanamine, 4-methyl,p-methylbenzylamine,p-tolylmethanamine,4-methylphenyl methanamine,benzylamine, p-methyl,p-xylylamine,4-methybenzylamine,1-4-methylphenyl methanamine,4-methyl-benzylamine
66035
(4-methylphenyl)methanamine
CC1=CC=C(C=C1)CN
104-84-7
C8H11N
121.18
MFCD00008123
HMTSWYPNXFHGEP-UHFFFAOYSA-N
4-methylbenzylamine,benzenemethanamine, 4-methyl,p-methylbenzylamine,p-tolylmethanamine,4-methylphenyl methanamine,benzylamine, p-methyl,p-xylylamine,4-methybenzylamine,1-4-methylphenyl methanamine,4-methyl-benzylamine
66035
(4-methylphenyl)methanamine
CC1=CC=C(C=C1)CN
Melting Point 13°C
Boiling Point 195°C
Specific Gravity 0.95
Flash Point 75°C
Refractive Index 1.5330 to 1.536
Packaging Glass bottle
Quantity 100g
Color Colorless to Yellow
Density 0.9500g/mL
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula CH3C6H4CH2NH2
Beilstein 12, 1141
Solubility Information Solubility in water: slightly soluble
Formula Weight 121.18
Physical Form Liquid
Percent Purity ≥97.5%
Chemical Name or Material 4-Methylbenzylamine, 98%
Show More Show Less

GHS P Statement Causes severe skin burns and eye damage.

GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.

GHS Signal Word: Danger

EINECSNumber : 203-243-2

TSCA : TSCA