Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methylcyclohexanone, 98%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
589-92-4
C7H12O
112.17
MFCD00001643
VGVHNLRUAMRIEW-UHFFFAOYSA-N
4-methylcyclohexanone,cyclohexanone, 4-methyl,4-methyl-1-cyclohexanone,methyl-4 cyclohexanone-1,p-methyl cyclohexanone,methycyclohexanone,metylocykloheksanon,unii-0l1r78k79s,4-methyl cyclohexanone,methyl-4 cyclohexanone-1 french
11525
4-methylcyclohexan-1-one
CC1CCC(=O)CC1
589-92-4
C7H12O
112.17
MFCD00001643
VGVHNLRUAMRIEW-UHFFFAOYSA-N
4-methylcyclohexanone,cyclohexanone, 4-methyl,4-methyl-1-cyclohexanone,methyl-4 cyclohexanone-1,p-methyl cyclohexanone,methycyclohexanone,metylocykloheksanon,unii-0l1r78k79s,4-methyl cyclohexanone,methyl-4 cyclohexanone-1 french
11525
4-methylcyclohexan-1-one
CC1CCC(=O)CC1
Infrared Spectrum Authentic
Packaging Glass Bottle
Specific Gravity 0.914
Melting Point -41°C
Flash Point 40°C
Boiling Point 169°C to 171°C
Refractive Index 1.4445 to 1.4465
Density 0.9140g/mL
Quantity 100g
Color Colorless to Yellow
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula CH3C6H9(=O)
Beilstein 07,18
Formula Weight 112.17
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 4-Methylcyclohexanone, 98%
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 209-665-3

RTECSNumber : GW1750200

TSCA : TSCA