Alfa Aesar™ 4-Methyldiphenylamine, 98%

Manufacturer: Alfa Aesar™ 4456018

Quantity 50g

 View more versions of this product

Catalog No. AA4456018

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 620-84-8
    Molecular Formula C13H13N
    Formula Weight 183.25
    MDL Number MFCD00092921
    Synonym N-Phenyl-p-toluidine
    Chemical Name or Material 4-Methyldiphenylamine

    Specifications

    Formula Weight 183.25
    Quantity 50g
    Molecular Formula C13H13N
    Health Hazard 1 Warning
    Flash Point 155°C (311°F)
    Color Pale brown
    Physical Form Crystalline
    CAS 620-84-8
    Health Hazard 3 GHS P Statement
    P280h-P305+P351+P338
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Boiling Point 334°C
    TSCA TSCA
    MDL Number MFCD00092921
    Synonym N-Phenyl-p-toluidine
    Health Hazard 2 GHS H Statement
    H302-H312-H315-H319
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    Melting Point 85° to 86°C
    Chemical Name or Material 4-Methyldiphenylamine
    Assay 98%

    GHS H Statement
    H302-H312-H315-H319
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P280h-P305+P351+P338
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit