Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methylmorpholine, 99%, Alfa Aesar™

Manufacturer:  Alfa Aesar A121580F

Quantity mL

 View more versions of this product

Catalog No. AAA121580F


  Check Availability
Add to cart

Specifications

Description

Applications
4-Methylmorpholine is used as a solvent for dyes, resins, waxes and pharmaceuticals. It acts as a crosslinker in the preparation of polyurethane foams, elastomers and adhesives. It is used as a precursor to prepare N-methylmorpholine N-oxide and morpholinium cation based ionic liquids. It is utilized as corrosion inhibitors and anti-scaling agents in industries.

Solubility
Miscible with water.

Notes
Incompatible with acids, acid chlorides, carbon dioxide and strong oxidizing agents.
Description & Specifications

Chemical Identifiers

109-02-4
101.15
UN2535
14,6277
4-Methylmorpholine
C5H11NO
MFCD00006175
102719
N-Methylmorpholine; NMM

Specifications

109-02-4
101.15
UN2535
14,6277
116° to 118°C
10.6
14°C (57°F)
4-Methylmorpholine
C5H11NO
MFCD00006175
102719
N-Methylmorpholine; NMM
−65°C
mL
Amine-like
99%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H225-H314-H318-H302-H312-H332;
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a;
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapours/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: 4-METHYLMORPHOLINE

RTECSNumber : QE5775000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit