4-Methylmorpholine, 99%, Alfa Aesar™

Manufacturer: Alfa Aesar A12158AE

Quantity 100mL

 View more versions of this product

Catalog No. AAA12158AE

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 109-02-4
    Physical Form Liquid
    Assay Percent Range 99%
    Molecular Formula C5H11NO
    Formula Weight 101.15
    Chemical Name or Material 4-Methylmorpholine

    Specifications

    Formula Weight 101.15
    Quantity 100mL
    Physical Form Liquid
    Density 0.920
    Assay Percent Range 99%
    Molecular Formula C5H11NO
    CAS 109-02-4
    Chemical Name or Material 4-Methylmorpholine

    Description

    Applications
    4-Methylmorpholine is used as a solvent for dyes, resins, waxes and pharmaceuticals. It acts as a crosslinker in the preparation of polyurethane foams, elastomers and adhesives. It is used as a precursor to prepare N-methylmorpholine N-oxide and morpholinium cation based ionic liquids. It is utilized as corrosion inhibitors and anti-scaling agents in industries.

    Solubility
    Miscible with water.

    Notes
    Incompatible with acids, acid chlorides, carbon dioxide and strong oxidizing agents.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit