Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
4-Methylmorpholine N-oxide, 50 wt.% solution in water, ACROS Organics™
Click to view available options
Quantity:
25g
100g
1kg
Packaging:
Glass bottle
Description
Oxidation reagent
Specifications
Packaging | Glass bottle |
Quantity | 25g |
Density | 1.1300g/mL |
Color | Colorless to Yellow |
Specific Gravity | 1.13 |
Flash Point | >100°C |
Boiling Point | 118.0°C to 119.0°C |
Melting Point | -20.0°C |
CAS Min % | 50.0,48.0 |
CAS Max % | 52.0,50.0 |
Show More |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Wear protective gloves/protective clothing/eye protec
GHS Signal Word: Warning
EINECSNumber : 231-391-8