Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Methylnicotinic acid hydrochloride, 97%

Click to view available options
Quantity:
0.25g
1g
5g
94015-05-1
C7H8ClNO2
173.596
MFCD00832700
QNKFCIXWPNMUAH-UHFFFAOYSA-N
4-methylnicotinic acid hydrochloride,4-methylpyridine-3-carboxylic acid hydrochloride,4-methylnicotinic acid hcl,4-methyl-nicotinic acid hydrochloride,acmc-209wrr,4-methyl nicotinic acid hydrochloride,4-methylnicotinic acid hydrochloride;,4-methylnicotinic acid monohydrochloride,4-methylnicotinic acid hydrochloride salt,4-methylnicotinic acid monohydrochloride;
2796506
4-methylpyridine-3-carboxylic acid;hydrochloride
CC1=C(C=NC=C1)C(=O)O.Cl
94015-05-1
C7H8ClNO2
173.596
MFCD00832700
QNKFCIXWPNMUAH-UHFFFAOYSA-N
4-methylnicotinic acid hydrochloride,4-methylpyridine-3-carboxylic acid hydrochloride,4-methylnicotinic acid hcl,4-methyl-nicotinic acid hydrochloride,acmc-209wrr,4-methyl nicotinic acid hydrochloride,4-methylnicotinic acid hydrochloride;,4-methylnicotinic acid monohydrochloride,4-methylnicotinic acid hydrochloride salt,4-methylnicotinic acid monohydrochloride;
2796506
4-methylpyridine-3-carboxylic acid;hydrochloride
CC1=C(C=NC=C1)C(=O)O.Cl
Quantity 5g
Formula Weight 173.6
Percent Purity 97%
Chemical Name or Material 4-Methylnicotinic acid hydrochloride

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning