Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Methyloctanoic acid, 98%

Click to view available options
Quantity:
5g
25g
54947-74-9
C9H18O2
158.241
MFCD00051938
LEGGANXCVQPIAI-UHFFFAOYSA-N
octanoic acid, 4-methyl,4-methyl-octanoic acid,4-methylcaprylic acid,+/--4-methyloctanoic acid,4-methyl-n-octanoic acid,4-methyl octanoic acid,octanoic acid, 4-methyl-, r,fema no. 3575,acmc-20mssw,acmc-20mssx
62089
4-methyloctanoic acid
CCCCC(C)CCC(=O)O
54947-74-9
C9H18O2
158.241
MFCD00051938
LEGGANXCVQPIAI-UHFFFAOYSA-N
octanoic acid, 4-methyl,4-methyl-octanoic acid,4-methylcaprylic acid,+/--4-methyloctanoic acid,4-methyl-n-octanoic acid,4-methyl octanoic acid,octanoic acid, 4-methyl-, r,fema no. 3575,acmc-20mssw,acmc-20mssx
62089
4-methyloctanoic acid
CCCCC(C)CCC(=O)O
Quantity 5g
Boiling Point 272°C
Flash Point >110°C (230°F)
UN Number UN3265
Beilstein 1755426
Formula Weight 158.24
Percent Purity 98%
Chemical Name or Material 4-Methyloctanoic acid

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

TSCA : TSCA