Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Methyloxindole, 95%

Click to view available options
Quantity:
250mg
1g
13220-46-7
C9H9NO
147.177
MFCD09835631
USRZZPHRQZGXFH-UHFFFAOYSA-N
4-methylindolin-2-one,4-methyloxindole,4-methyl-2-oxo-1,2-dihydro-indole,2h-indol-2-one, 1,3-dihydro-4-methyl,4-methylindolinon,4-methyl-oxindole,4-methyl-2-oxindole,4-methyl-1,3-dihydro-indol-2-one,4-methyl-1,3-dihydro-indole-2-one,4-methyl-2,3-dihydro-1h-indol-2-one
12579382
4-methyl-1,3-dihydroindol-2-one
CC1=C2CC(=O)NC2=CC=C1
13220-46-7
C9H9NO
147.177
MFCD09835631
USRZZPHRQZGXFH-UHFFFAOYSA-N
4-methylindolin-2-one,4-methyloxindole,4-methyl-2-oxo-1,2-dihydro-indole,2h-indol-2-one, 1,3-dihydro-4-methyl,4-methylindolinon,4-methyl-oxindole,4-methyl-2-oxindole,4-methyl-1,3-dihydro-indol-2-one,4-methyl-1,3-dihydro-indole-2-one,4-methyl-2,3-dihydro-1h-indol-2-one
12579382
4-methyl-1,3-dihydroindol-2-one
CC1=C2CC(=O)NC2=CC=C1
Quantity 250mg
Beilstein 119249
Formula Weight 147.18
Physical Form Solid
Percent Purity 95%
Chemical Name or Material 4-Methyloxindole