Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(4-Methylphenoxy)acetic acid, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
940-64-7
C9H10O3
166.18
MFCD00014365
SFTDDFBJWUWKMN-UHFFFAOYSA-N
4-methylphenoxy acetic acid,4-methylphenoxyacetic acid,2-4-methylphenoxy acetic acid,p-methylphenoxyacetic acid,unii-wwy23322ik,acetic acid, 4-methylphenoxy,2-p-tolyloxy acetic acid,p-toloxyacetic acid,acmc-20apmz,p-tolyloxy-acetic acid
70329
2-(4-methylphenoxy)acetic acid
CC1=CC=C(C=C1)OCC(=O)O
940-64-7
C9H10O3
166.18
MFCD00014365
SFTDDFBJWUWKMN-UHFFFAOYSA-N
4-methylphenoxy acetic acid,4-methylphenoxyacetic acid,2-4-methylphenoxy acetic acid,p-methylphenoxyacetic acid,unii-wwy23322ik,acetic acid, 4-methylphenoxy,2-p-tolyloxy acetic acid,p-toloxyacetic acid,acmc-20apmz,p-tolyloxy-acetic acid
70329
2-(4-methylphenoxy)acetic acid
CC1=CC=C(C=C1)OCC(=O)O
Melting Point 140°C to 142°C
Quantity 5g
Color Beige
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula CH3(C6H4)OCH2CO2H
Formula Weight 166.18
Physical Form Crystalline Powder
Percent Purity ≥98.5%
Chemical Name or Material (4-Methylphenoxy)acetic acid
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.