Alfa Aesar™ 4-(Methylphenylthio)acetaldehyde diethyl acetal, 98%

Manufacturer: Alfa Aesar™ A1177022

Quantity 100g

 View more versions of this product

Catalog No. AAA1177022

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 51830-50-3
    Molecular Formula C13H20O2S
    Formula Weight 240.37
    MDL Number MFCD00051603
    Beilstein 1964544
    Synonym p-(Tolylthio)acetaldehyde diethyl acetal
    Chemical Name or Material 4- (Methylphenylthio)acetaldehyde diethyl acetal

    Specifications

    Formula Weight 240.37
    MDL Number MFCD00051603
    CAS 51830-50-3
    Synonym p-(Tolylthio)acetaldehyde diethyl acetal
    Molecular Formula C13H20O2S
    Beilstein 1964544
    Boiling Point 106° to 108°C (0.25mmHg)
    Quantity 100g
    Chemical Name or Material 4- (Methylphenylthio)acetaldehyde diethyl acetal
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit