Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methylpiperidine, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
100g
500g
2.5kg
626-58-4
C6H13N
99.177
MFCD00006005
UZOFELREXGAFOI-UHFFFAOYSA-N
4-pipecoline,piperidine, 4-methyl,gamma-pipecoline,4-methylpiperidin,.gamma.-pipecoline,4-methyl-piperidine,4-methylpiperdine,4-methylpyperidine,4-methyl piperidine,hexahydro-g-picoline
69381
4-methylpiperidine
CC1CCNCC1
626-58-4
C6H13N
99.177
MFCD00006005
UZOFELREXGAFOI-UHFFFAOYSA-N
4-pipecoline,piperidine, 4-methyl,gamma-pipecoline,4-methylpiperidin,.gamma.-pipecoline,4-methyl-piperidine,4-methylpiperdine,4-methylpyperidine,4-methyl piperidine,hexahydro-g-picoline
69381
4-methylpiperidine
CC1CCNCC1
Quantity 5g
Color Yellow
Infrared Spectrum Authentic
Packaging Glass bottle
Density 0.8400g/mL
Refractive Index 1.4448 to 1.4468 (20°C, 589nm)
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Formula Weight 99.18
Physical Form Liquid
Percent Purity ≥98.5%
Chemical Name or Material 4-Methylpiperidine, 99%
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.
Causes serious eye damage.
Highly flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 210-954-1

TSCA : TSCA