Alfa Aesar™ 4'-(Methylsulfonyl)acetophenone, 97+%

Manufacturer: Alfa Aesar™ H5407203

Quantity 1g

 View more versions of this product

Catalog No. AAH5407203

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 10297-73-1
    Molecular Formula C9H10O3S
    Formula Weight 198.24
    MDL Number MFCD00025072
    Chemical Name or Material 4'-(Methylsulfonyl)acetophenone

    Specifications

    Formula Weight 198.24
    Molecular Formula C9H10O3S
    MDL Number MFCD00025072
    CAS 10297-73-1
    Melting Point 126° to 129°C
    Quantity 1g
    Chemical Name or Material 4'-(Methylsulfonyl)acetophenone
    Assay ≥97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit