Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Methylsulfonyl)aniline, ≥97%, Thermo Scientific™

Catalog Number AAH6469722
Click to view available options
Quantity:
25 g
100 g
500 g
5470-49-5
C7H9NO2S
171.21
MFCD00025353
XJEVFFNOMKXBLU-UHFFFAOYSA-N
4-methylsulfonyl aniline, 4-methylsulphonyl aniline, 4-mesylaniline, 4-methanesulfonylaniline, p-methylsulfonyl aniline, benzeneamine, 4-methylsulfonyl, benzenamine, 4-methylsulfonyl, 4-methylsulfonyl benzenamine, 4-methylsulphonylaniline
79617
CS(=O)(=O)C1=CC=C(N)C=C1
5470-49-5
C7H9NO2S
171.21
MFCD00025353
XJEVFFNOMKXBLU-UHFFFAOYSA-N
4-methylsulfonyl aniline, 4-methylsulphonyl aniline, 4-mesylaniline, 4-methanesulfonylaniline, p-methylsulfonyl aniline, benzeneamine, 4-methylsulfonyl, benzenamine, 4-methylsulfonyl, 4-methylsulfonyl benzenamine, 4-methylsulphonylaniline
79617
CS(=O)(=O)C1=CC=C(N)C=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is used as pharmaceutical intermediate. A series of 4-(methylsulfonyl)aniline derivatives were synthesized in order to obtain new compounds as a potential anti-inflammatory agents with expected selectivity against COX-2 enzyme.

Solubility
Soluble in water.

Notes
Light sensitive. Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 133°C to 138°C
Color White
Density 1.44 g/mL
Quantity 100 g
UN Number UN2811
Beilstein 2208425
Sensitivity Light sensitive
Solubility Information Soluble in water.
IUPAC Name 4-methylsulfonylaniline
Formula Weight 171.21
Percent Purity ≥97%
Physical Form Crystalline Powder
Chemical Name or Material 4-(Methylsulfonyl)aniline
Show More Show Less
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 226-803-8
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit