Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-(Methylsulfonyl)aniline, ≥97%

Click to view available options
Quantity:
25g
100g
500g
5470-49-5
C7H9NO2S
171.214
MFCD00025353
XJEVFFNOMKXBLU-UHFFFAOYSA-N
4-methylsulfonyl aniline,4-methylsulphonyl aniline,4-mesylaniline,4-methanesulfonylaniline,p-methylsulfonyl aniline,benzeneamine, 4-methylsulfonyl,4-methylsulfonyl aniline,benzenamine, 4-methylsulfonyl,4-methylsulfonyl benzenamine,4-methylsulphonylaniline
79617
4-methylsulfonylaniline
CS(=O)(=O)C1=CC=C(C=C1)N
5470-49-5
C7H9NO2S
171.214
MFCD00025353
XJEVFFNOMKXBLU-UHFFFAOYSA-N
4-methylsulfonyl aniline,4-methylsulphonyl aniline,4-mesylaniline,4-methanesulfonylaniline,p-methylsulfonyl aniline,benzeneamine, 4-methylsulfonyl,4-methylsulfonyl aniline,benzenamine, 4-methylsulfonyl,4-methylsulfonyl benzenamine,4-methylsulphonylaniline
79617
4-methylsulfonylaniline
CS(=O)(=O)C1=CC=C(C=C1)N
Applications
It is used as pharmaceutical intermediate. A series of 4-(methylsulfonyl)aniline derivatives were synthesized in order to obtain new compounds as a potential anti-inflammatory agents with expected selectivity against COX-2 enzyme.

Solubility
Soluble in water.

Notes
Light sensitive. Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
Sensitivity Light Sensitive
Quantity 500g
Melting Point 133°C to 138°C
Density 1.44g/mL
Color White
UN Number UN2811
Beilstein 2208425
Formula Weight 171.21
Physical Form Crystalline Powder
Percent Purity ≥97%
Chemical Name or Material 4-(Methylsulfonyl)aniline
Show More Show Less

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

storageNote1 : 2° to 8°C