Alfa Aesar™ 4-(Methylsulfonyl)phenol, 97%

Manufacturer: Alfa Aesar™ H3293509

Quantity 10g

 View more versions of this product

Catalog No. AAH3293509

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 14763-60-1
    Molecular Formula C7H8O3S
    Formula Weight 172.2
    MDL Number MFCD00047806
    Synonym 4-Hydroxyphenyl methyl sulfone
    Chemical Name or Material 4-(Methylsulfonyl)phenol

    Specifications

    Formula Weight 172.2
    Molecular Formula C7H8O3S
    MDL Number MFCD00047806
    CAS 14763-60-1
    Synonym 4-Hydroxyphenyl methyl sulfone
    Health Hazard 1 Warning
    Melting Point 90° to 95°C
    Quantity 10g
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    RTECS Number SM1530000
    Chemical Name or Material 4-(Methylsulfonyl)phenol
    Assay 97%

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit