Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methylthiazole-5-carboxaldehyde, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
82294-70-0
C5H5NOS
127.17
MFCD07368277
JJVIEMFQPALZOZ-UHFFFAOYSA-N
4-methylthiazole-5-carbaldehyde,4-methyl-5-thiazolecarboxaldehyde,4-methylthiazole-5-carboxaldehyde,4-methyl-5-formylthiazole,4-methyl-5-thiazolecarbaldehyde,5-formyl-4-methylthiazole,4-methylthiazole-5-aldehyde,4-methyl-5-thiazolyl aldehyde,5-thiazolecarboxaldehyde, 4-methyl,4-methyl-5-thiazole carboxaldehyde
581339
4-methyl-1,3-thiazole-5-carbaldehyde
CC1=C(SC=N1)C=O
82294-70-0
C5H5NOS
127.17
MFCD07368277
JJVIEMFQPALZOZ-UHFFFAOYSA-N
4-methylthiazole-5-carbaldehyde,4-methyl-5-thiazolecarboxaldehyde,4-methylthiazole-5-carboxaldehyde,4-methyl-5-formylthiazole,4-methyl-5-thiazolecarbaldehyde,5-formyl-4-methylthiazole,4-methylthiazole-5-aldehyde,4-methyl-5-thiazolyl aldehyde,5-thiazolecarboxaldehyde, 4-methyl,4-methyl-5-thiazole carboxaldehyde
581339
4-methyl-1,3-thiazole-5-carbaldehyde
CC1=C(SC=N1)C=O
Quantity 1g
Melting Point 74.0°C to 76.0°C
Packaging Glass bottle
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Formula Weight 127.17
Percent Purity 97%
Chemical Name or Material 4-Methylthiazole-5-carboxaldehyde

GHS H Statement
Causes skin irritation.
Causes serious eye damage.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately

GHS Signal Word: Danger