Alfa Aesar™ 4-Methylthiazole-5-carboxylic acid, 97%

Manufacturer: Alfa Aesar™ H3266209

Quantity 10g

 View more versions of this product

Catalog No. AAH3266209

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 20485-41-0
    Molecular Formula C5H5NO2S
    Formula Weight 143.17
    MDL Number MFCD00626872
    Chemical Name or Material 4-Methylthiazole-5-carboxylic acid

    Specifications

    Formula Weight 143.17
    Molecular Formula C5H5NO2S
    MDL Number MFCD00626872
    Health Hazard 1 Warning
    Melting Point 287°C
    Quantity 10g
    Health Hazard 2 GHS H Statement
    H317
    May cause an allergic skin reaction.
    Health Hazard 3 GHS P Statement
    P261-P280-P363-P321-P333+P313-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Wash contaminated clothing before reuse.
    Specific treatment (see label).
    If skin irritation or rash occurs: Get medical advice/attention.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 20485-41-0
    Chemical Name or Material 4-Methylthiazole-5-carboxylic acid
    Assay 97%

    GHS H Statement
    H317
    May cause an allergic skin reaction.

    GHS P Statement
    P261-P280-P363-P321-P333+P313-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Wash contaminated clothing before reuse.
    Specific treatment (see label).
    If skin irritation or rash occurs: Get medical advice/attention.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit