Alfa Aesar™ 4-Methylthio-2-butanone, 98%

Manufacturer: Alfa Aesar™ B2220614

Quantity 25g

 View more versions of this product

Catalog No. AAB2220614

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 34047-39-7
    Molecular Formula CH3SCH2CH2COCH3
    Formula Weight 118.2
    MDL Number MFCD00040029
    Synonym 4-(Methylmercapto)-2-butanone
    Chemical Name or Material 4-Methylthio-2-butanone

    Specifications

    Formula Weight 118.2
    MDL Number MFCD00040029
    Health Hazard 2 GHS H Statement
    H227
    Combustible liquid.
    Health Hazard 3 GHS P Statement
    P210-P280-P370+P378a-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    In case of fire: Use for extinction: CO2, powder or water spray.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Flash Point 70°C (158°F)
    Quantity 25g
    TSCA TSCA
    Molecular Formula CH3SCH2CH2COCH3
    CAS 34047-39-7
    Synonym 4-(Methylmercapto)-2-butanone
    Health Hazard 1 Warning
    Boiling Point 105° to 107°C (55mmHg)
    Chemical Name or Material 4-Methylthio-2-butanone
    Assay 98%

    GHS H Statement
    H227
    Combustible liquid.

    GHS P Statement
    P210-P280-P370+P378a-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    In case of fire: Use for extinction: CO2, powder or water spray.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit