Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Methylthio-2-butanone, 98%

Click to view available options
Quantity:
5g
25g
34047-39-7
C5H10OS
118.194
MFCD00040029
DRGHCRKOWMAZAO-UHFFFAOYSA-N
4-methylthio-2-butanone,2-butanone, 4-methylthio,4-methylthio butan-2-one,unii-5703w1o1ab,4-methylsulfanyl butan-2-one,potato butanone,fema no. 3375,5-thiahexane-2-one,acmc-1afub
61922
4-methylsulfanylbutan-2-one
CC(=O)CCSC
34047-39-7
C5H10OS
118.194
MFCD00040029
DRGHCRKOWMAZAO-UHFFFAOYSA-N
4-methylthio-2-butanone,2-butanone, 4-methylthio,4-methylthio butan-2-one,unii-5703w1o1ab,4-methylsulfanyl butan-2-one,potato butanone,fema no. 3375,5-thiahexane-2-one,acmc-1afub
61922
4-methylsulfanylbutan-2-one
CC(=O)CCSC
Quantity 5g
Flash Point 70°C (158°F)
Boiling Point 105°C to 107°C (55mmHg)
Formula Weight 118.2
Percent Purity 98%
Chemical Name or Material 4-Methylthio-2-butanone

GHS H Statement
H227
Combustible liquid.

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA