Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-(Methylthio)benzeneboronic acid, 97%

Click to view available options
Quantity:
1g
5g
25g
98546-51-1
C7H9BO2S
168.017
MFCD00093410
IVUHTLFKBDDICS-UHFFFAOYSA-N
4-methylthio phenylboronic acid,4-thioanisoleboronic acid,4-methylsulfanyl phenylboronic acid,4-methylthiophenylboronic acid,4-methylthio phenyl boronic acid,4-methylsulfanylphenyl boronic acid,4-methylthio phenylboronicacid,4-methylthio benzeneboronic acid,thioanisole-4-boronic acid,4-methylmercapto benzeneboronic acid
2733959
(4-methylsulfanylphenyl)boronic acid
B(C1=CC=C(C=C1)SC)(O)O
98546-51-1
C7H9BO2S
168.017
MFCD00093410
IVUHTLFKBDDICS-UHFFFAOYSA-N
4-methylthio phenylboronic acid,4-thioanisoleboronic acid,4-methylsulfanyl phenylboronic acid,4-methylthiophenylboronic acid,4-methylthio phenyl boronic acid,4-methylsulfanylphenyl boronic acid,4-methylthio phenylboronicacid,4-methylthio benzeneboronic acid,thioanisole-4-boronic acid,4-methylmercapto benzeneboronic acid
2733959
(4-methylsulfanylphenyl)boronic acid
B(C1=CC=C(C=C1)SC)(O)O
Quantity 1g
Melting Point ~208°C to 212°C
Beilstein 3247219
Formula Weight 168.02
Percent Purity 97%
Chemical Name or Material 4-(Methylthio)benzeneboronic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning