Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-(Methylthio)benzyl alcohol, 98%

Click to view available options
Quantity:
5g
25g
100g
3446-90-0
C8H10OS
154.227
MFCD00009706
MTXQKSQYMREAGJ-UHFFFAOYSA-N
4-methylthio benzyl alcohol,4-methylthio phenyl methanol,p-methylthiobenzyl alcohol,4-methylsulfanyl phenyl methanol,4-methylthiobenzyl alcohol,4-methylthio benzylalcohol,p-methylthio benzyl alcohol,4-methylsulfanylphenyl methanol,4-hydroxymethyl thioanisole
592968
(4-methylsulfanylphenyl)methanol
CSC1=CC=C(C=C1)CO
3446-90-0
C8H10OS
154.227
MFCD00009706
MTXQKSQYMREAGJ-UHFFFAOYSA-N
4-methylthio benzyl alcohol,4-methylthio phenyl methanol,p-methylthiobenzyl alcohol,4-methylsulfanyl phenyl methanol,4-methylthiobenzyl alcohol,4-methylthio benzylalcohol,p-methylthio benzyl alcohol,4-methylsulfanylphenyl methanol,4-hydroxymethyl thioanisole
592968
(4-methylsulfanylphenyl)methanol
CSC1=CC=C(C=C1)CO
Applications
4-(Methylthio)benzenemethanol is used to prepare (4-substituted benzyl)(trifluoromethyl)pyrazoles and -pyrazolones as potent antihyperglycemic agents.

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Quantity 100g
Boiling Point 108°C to 109°C (0.4mmHg)
Melting Point 41°C to 43°C
Flash Point >110°C (230°F)
Formula Weight 154.23
Percent Purity 98%
Chemical Name or Material 4-(Methylthio)benzyl alcohol