Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Methylthio)benzyl alcohol, 98%, ACROS Organics™

Click to view available options
Quantity:
10g
3446-90-0
C8H10OS
154.227
MFCD00009706
MTXQKSQYMREAGJ-UHFFFAOYSA-N
4-methylthio benzyl alcohol,4-methylthio phenyl methanol,p-methylthiobenzyl alcohol,4-methylsulfanyl phenyl methanol,4-methylthiobenzyl alcohol,4-methylthio benzylalcohol,p-methylthio benzyl alcohol,4-methylsulfanylphenyl methanol,4-hydroxymethyl thioanisole
592968
(4-methylsulfanylphenyl)methanol
CSC1=CC=C(C=C1)CO
3446-90-0
C8H10OS
154.227
MFCD00009706
MTXQKSQYMREAGJ-UHFFFAOYSA-N
4-methylthio benzyl alcohol,4-methylthio phenyl methanol,p-methylthiobenzyl alcohol,4-methylsulfanyl phenyl methanol,4-methylthiobenzyl alcohol,4-methylthio benzylalcohol,p-methylthio benzyl alcohol,4-methylsulfanylphenyl methanol,4-hydroxymethyl thioanisole
592968
(4-methylsulfanylphenyl)methanol
CSC1=CC=C(C=C1)CO
Infrared Spectrum Authentic
Quantity 10g
Color White to Yellow
Melting Point 38°C to 42°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula CH3SC6H4CH2OH
Beilstein 06, IV, 5919
Formula Weight 154.23
Physical Form Crystalline Powder
Percent Purity ≥97.5%
Chemical Name or Material 4-(Methylthio)benzyl alcohol, 98%
Show More Show Less