Alfa Aesar™ 4-(Methylthio)butanol, 99%

Manufacturer: Alfa Aesar™ B2267303

Quantity 1g

 View more versions of this product

Catalog No. AAB2267303

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 20582-85-8
    Molecular Formula C5H12OS
    Linear Formula CH3S(CH2)4OH
    Formula Weight 120.21
    MDL Number MFCD00002973
    Synonym 4-(Methylmercapto)butanol
    Chemical Name or Material 4-(Methylthio)butanol

    Specifications

    Formula Weight 120.21
    Flash Point 101°C (215°F)
    Quantity 1g
    Molecular Formula C5H12OS
    Linear Formula CH3S(CH2)4OH
    MDL Number MFCD00002973
    CAS 20582-85-8
    Synonym 4-(Methylmercapto)butanol
    Odor Unpleasant
    Boiling Point 95° to 98°C (20mmHg)
    Chemical Name or Material 4-(Methylthio)butanol
    Assay 99%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit