Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-(Methylthio)butanol, 99%

Click to view available options
Quantity:
1g
5g
25g
20582-85-8
C5H12OS
120.21
MFCD00002973
JNTVUHZXIJFHAU-UHFFFAOYSA-N
4-methylthio-1-butanol,4-methylthio butanol,4-methylthiobutan-1-ol,unii-yl87ury6pc,4-methylsulfanyl-1-butanol,1-butanol, 4-methylthio,yl87ury6pc,4-methylthio butan-1-ol,4-methylsulfanyl butan-1-ol,4-methylthio-1-butanol, polymer-bound
519793
4-methylsulfanylbutan-1-ol
CSCCCCO
20582-85-8
C5H12OS
120.21
MFCD00002973
JNTVUHZXIJFHAU-UHFFFAOYSA-N
4-methylthio-1-butanol,4-methylthio butanol,4-methylthiobutan-1-ol,unii-yl87ury6pc,4-methylsulfanyl-1-butanol,1-butanol, 4-methylthio,yl87ury6pc,4-methylthio butan-1-ol,4-methylsulfanyl butan-1-ol,4-methylthio-1-butanol, polymer-bound
519793
4-methylsulfanylbutan-1-ol
CSCCCCO
Odor Unpleasant
Boiling Point 95°C to 98°C (20mmHg)
Flash Point 101°C (215°F)
Quantity 25g
Linear Formula CH3S(CH2)4OH
Formula Weight 120.21
Percent Purity 99%
Chemical Name or Material 4-(Methylthio)butanol