Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Methylthio)phenylboronic acid, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass Bottle
98546-51-1
C7H9BO2S
168.017
MFCD00093410
IVUHTLFKBDDICS-UHFFFAOYSA-N
4-methylthio phenylboronic acid,4-thioanisoleboronic acid,4-methylsulfanyl phenylboronic acid,4-methylthiophenylboronic acid,4-methylthio phenyl boronic acid,4-methylsulfanylphenyl boronic acid,4-methylthio phenylboronicacid,4-methylthio benzeneboronic acid,thioanisole-4-boronic acid,4-methylmercapto benzeneboronic acid
2733959
(4-methylsulfanylphenyl)boronic acid
B(C1=CC=C(C=C1)SC)(O)O
98546-51-1
C7H9BO2S
168.017
MFCD00093410
IVUHTLFKBDDICS-UHFFFAOYSA-N
4-methylthio phenylboronic acid,4-thioanisoleboronic acid,4-methylsulfanyl phenylboronic acid,4-methylthiophenylboronic acid,4-methylthio phenyl boronic acid,4-methylsulfanylphenyl boronic acid,4-methylthio phenylboronicacid,4-methylthio benzeneboronic acid,thioanisole-4-boronic acid,4-methylmercapto benzeneboronic acid
2733959
(4-methylsulfanylphenyl)boronic acid
B(C1=CC=C(C=C1)SC)(O)O
Quantity 5g
Packaging Glass Bottle
Color White
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Formula Weight 168.02
Physical Form Crystalline Powder
Percent Purity ≥96.0%
Chemical Name or Material 4-(Methylsulfanyl)phenylboronic acid
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.