Alfa Aesar™ 4-Methylumbelliferyl acetate, 99%

Manufacturer: Alfa Aesar™ A1214722

Quantity 100g

 View more versions of this product

Catalog No. AAA1214722

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2747-05-9
    Molecular Formula C12H10O4
    Formula Weight 218.21
    MDL Number MFCD00006865
    Beilstein 189667
    Synonym 7-Acetoxy-4-methylcoumarin
    Chemical Name or Material 4-Methylumbelliferyl acetate

    Specifications

    Formula Weight 218.21
    Molecular Formula C12H10O4
    CAS 2747-05-9
    Beilstein 189667
    Synonym 7-Acetoxy-4-methylcoumarin
    Quantity 100g
    Recommended Storage 2°C to 8°C
    MDL Number MFCD00006865
    Melting Point 147° to 152°C
    Chemical Name or Material 4-Methylumbelliferyl acetate
    Assay 99%

    2°C to 8°C

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit