Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methylumbelliferyl-β-D-glucuronide dihydrate, 98%, Acros Organics™

Click to view available options
Quantity:
1g
5g
25g
6160-80-1
C16H16O9·2H2O
388.33
ARQXEQLMMNGFDU-JHZZJYKESA-N
4-methylumbelliferyl-beta-d-glucuronide,4-methylumbelliferyl glucuronide,4-methylumbelliferone glucuronide,mug trihydrate,4-methylumbelliferyl beta-glucuronide,4-methyl-2-oxo-2h-1-benzopyran-7-yl beta-d-glucopyranosiduronic acid,2s,3s,4s,5r,6s-3,4,5-trihydroxy-6-4-methyl-2-oxo-2h-chromen-7-yl oxy tetrahydro-2h-pyran-2-carboxylic acid,2s,3s,4s,5r,6s-3,4,5-trihydroxy-6-4-methyl-2-oxo-chromen-7-yl oxy-tetrahydropyran-2-carboxylic acid,2s,3s,4s,6s,5r-3,4,5-trihydroxy-6-4-methyl-2-oxochromen-7-yloxy-2h-3,4,5,6-tetrahydropyran-2-carboxylic acid,2s,3s,4s,5r,6s-3,4,5-trihydroxy-6-4-methyl-2-oxochromen-7-yl oxy oxane-2-carboxylic acid
91553
CHEBI:1904
(2S,3S,4S,5R,6S)-3,4,5-trihydroxy-6-(4-methyl-2-oxochromen-7-yl)oxyoxane-2-carboxylic acid
CC1=CC(=O)OC2=C1C=CC(=C2)OC3C(C(C(C(O3)C(=O)O)O)O)O
6160-80-1
C16H16O9·2H2O
388.33
ARQXEQLMMNGFDU-JHZZJYKESA-N
4-methylumbelliferyl-beta-d-glucuronide,4-methylumbelliferyl glucuronide,4-methylumbelliferone glucuronide,mug trihydrate,4-methylumbelliferyl beta-glucuronide,4-methyl-2-oxo-2h-1-benzopyran-7-yl beta-d-glucopyranosiduronic acid,2s,3s,4s,5r,6s-3,4,5-trihydroxy-6-4-methyl-2-oxo-2h-chromen-7-yl oxy tetrahydro-2h-pyran-2-carboxylic acid,2s,3s,4s,5r,6s-3,4,5-trihydroxy-6-4-methyl-2-oxo-chromen-7-yl oxy-tetrahydropyran-2-carboxylic acid,2s,3s,4s,6s,5r-3,4,5-trihydroxy-6-4-methyl-2-oxochromen-7-yloxy-2h-3,4,5,6-tetrahydropyran-2-carboxylic acid,2s,3s,4s,5r,6s-3,4,5-trihydroxy-6-4-methyl-2-oxochromen-7-yl oxy oxane-2-carboxylic acid
91553
CHEBI:1904
(2S,3S,4S,5R,6S)-3,4,5-trihydroxy-6-(4-methyl-2-oxochromen-7-yl)oxyoxane-2-carboxylic acid
CC1=CC(=O)OC2=C1C=CC(=C2)OC3C(C(C(C(O3)C(=O)O)O)O)O
Specific Rotation -105
Specific Rotation Condition −105.00 (30.00°C c=0.25, water)
Quantity 5g
Melting Point 102.0°C
Infrared Spectrum Authentic
Water 8 to 10.2% (K.F.)
Assay Percent Range 97.5% min. (HPLC)
Solubility Information (c=1,H2O at 60°C) Clear colorless solution
Formula Weight 388.33
Percent Purity 98%
Chemical Name or Material 4-Methylumbelliferyl-β-D-glucuronide dihydrate
Show More Show Less

EINECSNumber : 228-186-