Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methylumbelliferyl-N-acetyl-β-D-galactosaminide hydrate, 98%, ACROS Organics™

Click to view available options
Quantity:
250mg
Packaging:
Glass bottle
36476-29-6
C18H21NO8·xH2O
379.37
MFCD03791283
QCTHLCFVVACBSA-SOVHRIKKSA-M
22842632
N-[(2S,3R,4R,5R,6R)-4,5-dihydroxy-6-(hydroxymethyl)-2-(4-methyl-2-oxochromen-7-yl)oxyoxan-3-yl]ethanimidate
CC1=CC(=O)OC2=C1C=CC(=C2)OC3C(C(C(C(O3)CO)O)O)N=C(C)[O-]
36476-29-6
C18H21NO8·xH2O
379.37
MFCD03791283
QCTHLCFVVACBSA-SOVHRIKKSA-M
22842632
N-[(2S,3R,4R,5R,6R)-4,5-dihydroxy-6-(hydroxymethyl)-2-(4-methyl-2-oxochromen-7-yl)oxyoxan-3-yl]ethanimidate
CC1=CC(=O)OC2=C1C=CC(=C2)OC3C(C(C(C(O3)CO)O)O)N=C(C)[O-]
Appearance (1% soln. in warm methanol/water (1:1)) clear, colorless to faint yellow
Quantity 250mg
Water 3% max. (K.F.)
Color White to Yellow
Specific Rotation − 45.00
Melting Point 190.0°C to 192.0°C
Specific Rotation Condition − 45.00 (20.00°C c=0.3,pyridine)
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Solubility Information (1% in warm MeOH/water 1:1) Clear colorless to light yellow
Formula Weight 379.37
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material 4-Methylumbelliferyl-N-acetyl-β-D-galactosaminide hydrate, 98%
Show More Show Less

EINECSNumber : 253-052-3