Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methylumbelliferylphosphate disodium salt trihydrate, 99%, ACROS Organics™

Click to view available options
Quantity:
250mg
Packaging:
Glass bottle
22919-26-2
C10H7Na2O63H2O
354.16
MFCD03066085
WUUDJQVNZPEPKN-UHFFFAOYSA-L
sodium 4-methyl-2-oxo-2h-chromen-7-yl phosphate,2h-1-benzopyran-2-one, 4-methyl-7-phosphonooxy-, disodium salt,4-methylumbelliferyl phosphate disodium salt,4-methylumbelliferyl phosphate sodium salt,2h-1-benzopyran-2-one, 4-methyl-7-phosphonooxy-, sodium salt 1:2,disodium 4-methyl-2-oxochromen-7-yl phosphate,4-methylumbelliferyl phosphate, disodium salt,disodium 7-hydroxy-4-methylcoumarinyl phosphate,disodium 4-methyl-2-oxo-2h-chromen-7-yl phosphate,disodium 4-methylumbelliferyl phosphate
89903
disodium;(4-methyl-2-oxochromen-7-yl) phosphate
CC1=CC(=O)OC2=C1C=CC(=C2)OP(=O)([O-])[O-].[Na+].[Na+]
22919-26-2
C10H7Na2O63H2O
354.16
MFCD03066085
WUUDJQVNZPEPKN-UHFFFAOYSA-L
sodium 4-methyl-2-oxo-2h-chromen-7-yl phosphate,2h-1-benzopyran-2-one, 4-methyl-7-phosphonooxy-, disodium salt,4-methylumbelliferyl phosphate disodium salt,4-methylumbelliferyl phosphate sodium salt,2h-1-benzopyran-2-one, 4-methyl-7-phosphonooxy-, sodium salt 1:2,disodium 4-methyl-2-oxochromen-7-yl phosphate,4-methylumbelliferyl phosphate, disodium salt,disodium 7-hydroxy-4-methylcoumarinyl phosphate,disodium 4-methyl-2-oxo-2h-chromen-7-yl phosphate,disodium 4-methylumbelliferyl phosphate
89903
disodium;(4-methyl-2-oxochromen-7-yl) phosphate
CC1=CC(=O)OC2=C1C=CC(=C2)OP(=O)([O-])[O-].[Na+].[Na+]
Infrared Spectrum Authentic
Color White
Quantity 250mg
Water 14.26 to 16.26% (K.F.)
Packaging Glass bottle
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Solubility Information Solubility in water: soluble in water
Formula Weight 354.16
Physical Form Powder
Percent Purity 99%
Chemical Name or Material 4-Methylumbelliferylphosphate disodium salt trihydrate, 99%
Show More Show Less

MOISTURE SENSITIVE

LIGHT SENSITIVE

EINECSNumber : 245-325-