Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Morpholinoacetophenone 99%, ACROS Organics™

Click to view available options
Quantity:
5g
50g
Packaging:
Glass bottle
39910-98-0
C12H15NO2
205.26
MFCD00006168
AKQWEDMTPCAESO-UHFFFAOYSA-N
4'-morpholinoacetophenone,1-4-morpholinophenyl ethanone,4-morpholinoacetophenone,acetophenone, 4'-morpholino,ethanone, 1-4-4-morpholinyl phenyl,p-morpholinoacetophenone,1-4-morpholin-4-yl phenyl ethan-1-one,p-morpholino acetophenone,4;-morpholinoacetophenone,1-4-morpholin-4-ylphenyl ethanone
38362
1-(4-morpholin-4-ylphenyl)ethanone
CC(=O)C1=CC=C(C=C1)N2CCOCC2
39910-98-0
C12H15NO2
205.26
MFCD00006168
AKQWEDMTPCAESO-UHFFFAOYSA-N
4'-morpholinoacetophenone,1-4-morpholinophenyl ethanone,4-morpholinoacetophenone,acetophenone, 4'-morpholino,ethanone, 1-4-4-morpholinyl phenyl,p-morpholinoacetophenone,1-4-morpholin-4-yl phenyl ethan-1-one,p-morpholino acetophenone,4;-morpholinoacetophenone,1-4-morpholin-4-ylphenyl ethanone
38362
1-(4-morpholin-4-ylphenyl)ethanone
CC(=O)C1=CC=C(C=C1)N2CCOCC2
Melting Point 94°C to 98°C
Quantity 50g
Infrared Spectrum Authentic
Color Yellow to Brown
Packaging Glass bottle
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Formula Weight 205.26
Physical Form Crystalline Powder
Percent Purity ≥98.5%
Chemical Name or Material 4-Morpholinoacetophenone, 99%
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 254-698-9

RTECSNumber : AM9486200