Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Morpholinoaniline, 98+%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
2524-67-6
C10H14N2O
178.23
MFCD00006169
PHNDZBFLOPIMSM-UHFFFAOYSA-N
4-morpholinoaniline,4-morpholin-4-yl-phenylamine,4-morpholin-4-yl aniline,n-4-aminophenyl morpholine,p-morpholinoaniline,4-morpholinobenzenamine,4-4-morpholino aniline,benzenamine, 4-4-morpholinyl,4-morpholinyl aniline,4-4-aminophenyl-morpholine
75655
4-morpholin-4-ylaniline
C1COCCN1C2=CC=C(C=C2)N
2524-67-6
C10H14N2O
178.23
MFCD00006169
PHNDZBFLOPIMSM-UHFFFAOYSA-N
4-morpholinoaniline,4-morpholin-4-yl-phenylamine,4-morpholin-4-yl aniline,n-4-aminophenyl morpholine,p-morpholinoaniline,4-morpholinobenzenamine,4-4-morpholino aniline,benzenamine, 4-4-morpholinyl,4-morpholinyl aniline,4-4-aminophenyl-morpholine
75655
4-morpholin-4-ylaniline
C1COCCN1C2=CC=C(C=C2)N
Melting Point 130°C to 131°C
Infrared Spectrum Authentic
Packaging Glass bottle
Color Purple to Brown
Quantity 25g
CAS Min % 98.0
CAS Max % 100.0
Assay Percent Range 98+%
Formula Weight 178.23
Physical Form Crystalline Powder
Percent Purity 98+%
Chemical Name or Material 4-Morpholinoaniline
Show More Show Less

GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 219-760-1