Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Morpholinophenyl isothiocyanate, Thermo Scientific™

Catalog Number p-4510163
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
51317-66-9
C11H12N2OS
220.29
AXUXRZZYZBZQAR-UHFFFAOYSA-N
224862
4-(4-isothiocyanatophenyl)morpholine
C1COCCN1C2=CC=C(C=C2)N=C=S
51317-66-9
C11H12N2OS
220.29
AXUXRZZYZBZQAR-UHFFFAOYSA-N
224862
4-(4-isothiocyanatophenyl)morpholine
C1COCCN1C2=CC=C(C=C2)N=C=S
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Synonym 4-morpholinophenyl isothiocyanate, 4-4-isothiocyanatophenyl morpholine, 4-morpholinophenylisothiocyanate, morpholine, 4-4-isothiocyanatophenyl, 4-4-isothiocyanatophenyl morpholin, 4-morpholin-4-ylbenzenisothiocyanate, 4-4-morpholinyl phenyl isothiocyanate, 4-4-isothiocyanato-phenyl-morpholine, morpholine,4-4-isothiocyanatophenyl, 4-morpholin-4-yl phenyl isothiocyanate
Formula Weight 220.29
Percent Purity 97%
Chemical Name or Material 4-Morpholinophenyl isothiocyanate
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit