Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-N-BOC-4-N-methylaminopiperidine, 98%, Acros Organics™

Click to view available options
Quantity:
1g
5g
108612-54-0
C11H22N2O2
214.31
DJJOYDXRUBOZON-UHFFFAOYSA-N
tert-butyl methyl piperidin-4-yl carbamate,4-n-boc-4-n-methyl-aminopiperidine,4-n-boc-4-n-methylaminopiperidine,tert-butyl n-methyl-n-piperidin-4-yl carbamate,4-n-boc-n-methylamino piperidine,4-boc-4-methylaminopiperidine,methyl-piperidin-4-yl-carbamic acid tert-butyl ester,tert-butyl n-methyl-n-4-piperidyl carbamate,tert-butyl methylpiperidin-4-ylcarbamate,methyl-piperidine-4-yl-carbamic acid tert-butyl ester
2756806
tert-butyl N-methyl-N-piperidin-4-ylcarbamate
CC(C)(C)OC(=O)N(C)C1CCNCC1
108612-54-0
C11H22N2O2
214.31
DJJOYDXRUBOZON-UHFFFAOYSA-N
tert-butyl methyl piperidin-4-yl carbamate,4-n-boc-4-n-methyl-aminopiperidine,4-n-boc-4-n-methylaminopiperidine,tert-butyl n-methyl-n-piperidin-4-yl carbamate,4-n-boc-n-methylamino piperidine,4-boc-4-methylaminopiperidine,methyl-piperidin-4-yl-carbamic acid tert-butyl ester,tert-butyl n-methyl-n-4-piperidyl carbamate,tert-butyl methylpiperidin-4-ylcarbamate,methyl-piperidine-4-yl-carbamic acid tert-butyl ester
2756806
tert-butyl N-methyl-N-piperidin-4-ylcarbamate
CC(C)(C)OC(=O)N(C)C1CCNCC1
Packaging Glass Bottle
Melting Point 39.0°C to 41.0°C
Quantity 1g
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Formula Weight 214.31
Percent Purity 98%
Chemical Name or Material 4-N-BOC-4-N-methylaminopiperidine

GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Call a POISON CENTER or doctor/physician if you feel unwell.

GHS Signal Word: Warning