Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-N-BOC-Amino-4-carboxytetrahydropyran, 95%, ACROS Organics™

Click to view available options
Quantity:
500mg
2g
10g
Packaging:
Glass bottle
172843-97-9
C11H19NO5
245.27
MFCD02683136
SPPDKPRJPFTBEV-UHFFFAOYSA-N
4-boc-amino tetrahydropyran-4-carboxylic acid,4-n-boc-amino-4-carboxytetrahydropyran,4-tert-butoxycarbonyl amino tetrahydro-2h-pyran-4-carboxylic acid,4-tert-butoxycarbonyl amino oxane-4-carboxylic acid,4-tert-butoxy carbonyl amino oxane-4-carboxylic acid,4-tert-butoxycarbamoyl tetrahydropyran-4-carboxylic acid,4-tert-butoxycarbonylamino tetrahydro-2h-pyran-4-carboxylic acid,4-tert-butoxy carbonylamino-2h-3,4,5,6-tetrahydropyran-4-carboxylic acid
1268219
4-[(2-methylpropan-2-yl)oxycarbonylamino]oxane-4-carboxylic acid
CC(C)(C)OC(=O)NC1(CCOCC1)C(=O)O
172843-97-9
C11H19NO5
245.27
MFCD02683136
SPPDKPRJPFTBEV-UHFFFAOYSA-N
4-boc-amino tetrahydropyran-4-carboxylic acid,4-n-boc-amino-4-carboxytetrahydropyran,4-tert-butoxycarbonyl amino tetrahydro-2h-pyran-4-carboxylic acid,4-tert-butoxycarbonyl amino oxane-4-carboxylic acid,4-tert-butoxy carbonyl amino oxane-4-carboxylic acid,4-tert-butoxycarbamoyl tetrahydropyran-4-carboxylic acid,4-tert-butoxycarbonylamino tetrahydro-2h-pyran-4-carboxylic acid,4-tert-butoxy carbonylamino-2h-3,4,5,6-tetrahydropyran-4-carboxylic acid
1268219
4-[(2-methylpropan-2-yl)oxycarbonylamino]oxane-4-carboxylic acid
CC(C)(C)OC(=O)NC1(CCOCC1)C(=O)O
Infrared Spectrum Authentic
Color White
Packaging Glass bottle
Quantity 10g
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Formula Weight 245.27
Physical Form Powder
Percent Purity 95%
Chemical Name or Material 4-N-BOC-Amino-4-carboxytetrahydropyran, 95%
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning