Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-N-BOC-Aminopiperidine, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
73874-95-0
C10H20N2O2
200.28
MFCD00798171
CKXZPVPIDOJLLM-UHFFFAOYSA-N
4-boc-aminopiperidine,tert-butyl piperidin-4-ylcarbamate,4-n-boc-aminopiperidine,4-boc-amino piperidine,4-n-boc-amino piperidine,tert-butyl n-piperidin-4-yl carbamate,4-tert-butoxycarbonylamino piperidine,4-n-boc-amino-piperidine,4-boc-amino-piperidine,4-n-boc amino piperidine
723833
tert-butyl N-piperidin-4-ylcarbamate
CC(C)(C)OC(=O)NC1CCNCC1
73874-95-0
C10H20N2O2
200.28
MFCD00798171
CKXZPVPIDOJLLM-UHFFFAOYSA-N
4-boc-aminopiperidine,tert-butyl piperidin-4-ylcarbamate,4-n-boc-aminopiperidine,4-boc-amino piperidine,4-n-boc-amino piperidine,tert-butyl n-piperidin-4-yl carbamate,4-tert-butoxycarbonylamino piperidine,4-n-boc-amino-piperidine,4-boc-amino-piperidine,4-n-boc amino piperidine
723833
tert-butyl N-piperidin-4-ylcarbamate
CC(C)(C)OC(=O)NC1CCNCC1
Quantity 5g
Melting Point 162.0°C to 166.0°C
Color White
Infrared Spectrum Authentic
Packaging Glass bottle
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 200.28
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material 4-N-BOC-Aminopiperidine, 98%
Show More Show Less

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger