Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-n-Butyl-4'-cyanobiphenyl, 98%

Click to view available options
Quantity:
1g
5g
52709-83-8
C17H17N
235.33
MFCD00506009
PJPLBHHDTUICNN-UHFFFAOYSA-N
4'-butyl-1,1'-biphenyl-4-carbonitrile,4-butyl-4'-cyanobiphenyl,4'-butyl-4-biphenylcarbonitrile,4-cyano-4'-butylbiphenyl,1,1'-biphenyl-4-carbonitrile, 4'-butyl,4-n-butyl-4'-cyanobiphenyl,4-4-butylphenyl benzonitrile,4'-butyl 1,1'-biphenyl-4-carbonitrile,4'-butyl-4-cyanobiphenyl
104288
4-(4-butylphenyl)benzonitrile
CCCCC1=CC=C(C=C1)C2=CC=C(C=C2)C#N
52709-83-8
C17H17N
235.33
MFCD00506009
PJPLBHHDTUICNN-UHFFFAOYSA-N
4'-butyl-1,1'-biphenyl-4-carbonitrile,4-butyl-4'-cyanobiphenyl,4'-butyl-4-biphenylcarbonitrile,4-cyano-4'-butylbiphenyl,1,1'-biphenyl-4-carbonitrile, 4'-butyl,4-n-butyl-4'-cyanobiphenyl,4-4-butylphenyl benzonitrile,4'-butyl 1,1'-biphenyl-4-carbonitrile,4'-butyl-4-cyanobiphenyl
104288
4-(4-butylphenyl)benzonitrile
CCCCC1=CC=C(C=C1)C2=CC=C(C=C2)C#N
Quantity 5g
Melting Point 49°C to 51°C
Boiling Point 210°C to 212°C (2mmHg)
UN Number UN3439
Formula Weight 235.33
Percent Purity 98%
Chemical Name or Material 4-n-Butyl-4'-cyanobiphenyl

GHS H Statement
H331-H304-H312
Toxic if inhaled.
May be fatal if swallowed and enters airways.
Harmful in contact with skin.

GHS P Statement
P261-P280-P301+P310-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

TSCA : TSCA