Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4'-n-Butylacetophenone, 95%, ACROS Organics™

Click to view available options
Quantity:
5mL
Packaging:
Glass bottle
37920-25-5
C12H16O
176.26
MQESVSITPLILCO-UHFFFAOYSA-N
4'-butylacetophenone,1-4-butylphenyl ethanone,ethanone, 1-4-butylphenyl,p-butylacetophenone,1-4-butylphenyl ethan-1-one,p-n-butylacetophenone,4'-n-butylacetophenone,unii-6738cf9z8v,1-acetyl-4-butylbenzene,1-4-butyl-phenyl ethanone
123470
1-(4-butylphenyl)ethanone
CCCCC1=CC=C(C=C1)C(=O)C
37920-25-5
C12H16O
176.26
MQESVSITPLILCO-UHFFFAOYSA-N
4'-butylacetophenone,1-4-butylphenyl ethanone,ethanone, 1-4-butylphenyl,p-butylacetophenone,1-4-butylphenyl ethan-1-one,p-n-butylacetophenone,4'-n-butylacetophenone,unii-6738cf9z8v,1-acetyl-4-butylbenzene,1-4-butyl-phenyl ethanone
123470
1-(4-butylphenyl)ethanone
CCCCC1=CC=C(C=C1)C(=O)C
Specific Gravity 0.957
Boiling Point 101°C to 102°C (1.5mmHg)
Flash Point >113°C
Color Colorless to Yellow
Density 0.9570g/mL
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 5mL
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 94% min. (GC)
Formula Weight 176.26
Physical Form Liquid
Percent Purity 95%
Chemical Name or Material 4'-n-Butylacetophenone
Show More Show Less

EINECSNumber : 253-715-7